Learn More
CaMKII delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15443-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specific to the C-term of delta chain, CaMKII delta. It can recognize most isoforms the gene encodes, from 50-72 kDa. It can recognize isoforms 2, 3, 4, 9, 11.
CaMKII delta belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. is a delta chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patternsSpécification
CaMKII delta | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13557 | |
CaM kinase II subunit delta, CaMK II subunit delta, CAMK2D, CAMKD, CaMKII delta | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
108058, 24246, 817 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CAMK2D | |
Liquid | |
CAMK2D | |
CaMKII delta Fusion Protein Ag7323 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |