missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKIV Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | CaMKIV |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16801394
|
Proteintech
13263-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16891384
|
Proteintech
13263-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CaM kinase IV (CaMKIV), in a calcium and calmodulin dependent manner, is implicated in phosphorylating numerous transcription factors including the CCAAT enhancer binding protein (C/EBP), serum response factor (SRF), and cyclic AMP response element binding protein (CBP). The nuclear localization of this protein is consistent with its role in mediating calcium-dependent gene expression. CaMKIV is particularly abundant in testis, T-cells, and neurons but is also found in other tissues to varying degrees. In neurons, CaMKIV is thought to play an important role in synaptic plasticity via its gene regulatory effects. In T-cells, this protein plays an important role in calcium signaling which could affect the transcription regulatory protein, nuclear factor of activated T-cells (NFAT). CaMKIV is encoded, along with calspermin, by the CaMKIV gene. It has been found that, in testes, CaMKIV is expressed in germ cells and found to be associated with chromatin. The association of CaMKIV with chromatin suggests a potential role in chromatin remodeling during nuclear condensation in spermatids.Spécification
CaMKIV | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P08414, P13234, Q16566 | |
12326, 25050, 814 | |
CaMKIV Fusion Protein Ag3922 | |
Primary | |
-20°C | |
CAMK4 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CaM kinase GR, CaMK GR, CAMK4, CaMKIV | |
CAMK4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |