missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13731-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Carboxypeptidases have functions ranging from digestion of food to selective biosynthesis of neuroendocrine peptides. Members of the A/B subfamily of carboxypeptidases, such as CPA5, contain an approximately 90-amino acid pro region that assists in the folding of the active carboxypeptidase domain. Cleavage of the pro region activates the enzyme.Spécification
Carboxypeptidase A5 | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R4H4, Q8WXQ8 | |
Carboxypeptidase A5, CPA5 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
408212, 74649, 93979 |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
CPA5 | |
Liquid | |
Cpa5 | |
Carboxypeptidase A5 Fusion Protein Ag4755 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human, Rat |