missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase M Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16440-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16806064

  • 112.71€ / 20µL

Expédition estimée
04-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.
Spécification

Spécification

Carboxypeptidase M
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14384
Carboxypeptidase M, CPM
Rabbit
IgG
20 μL
-20°C
Primary
1368
Western Blot
Unconjugated
CPM
Liquid
CPM
Carboxypeptidase M Fusion Protein Ag9761
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents