missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARMIL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27133-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Cell membrane-cytoskeleton-associated protein that plays a role in the regulation of actin polymerization at the barbed end of actin filaments. Prevents F-actin heterodimeric capping protein (CP) activity at the leading edges of migrating cells, and hence generates uncapped barbed ends and enhances actin polymerization, however, seems unable to nucleate filaments. Plays a role in lamellipodial protrusion formations and cell migration.Spécification
CARMIL | |
Polyclonal | |
Unconjugated | |
CARMIL1 | |
CARMIL, CARMIL homolog, CARMIL1a, LRRC16, LRRC16A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55604 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VZK9 | |
CARMIL1 | |
CARMIL Fusion Protein Ag25876 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |