missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15619-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.Spécification
CBR3 | |
Polyclonal | |
Unconjugated | |
CBR3 | |
Carbonyl reductase [NADPH] 3, carbonyl reductase 3, CBR3, hCBR3, SDR21C2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109857, 304078, 874 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75828, Q8K354 | |
CBR3 | |
CBR3 Fusion Protein Ag8039 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |