missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CCK (Human) Recombinant Protein
Human CCK full-length ORF ( AAH08283, 1 a.a. - 115 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00000885-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: MNSGVCLCVLMAVLAAGALTQPVPPADPAGSGLQRAEEAPRRQLRVSQRTDGESRAHLGALLARYIQQARKAPSGRMSIVKNLQNLDPSHRISDRDYMGWMDFGRRSAEEYEYPS
Spécification
AAH08283 | |
cholecystokinin | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCK | |
GST |
885 | |
Wheat germ expression system | |
25 μg | |
MGC117187 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |