missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL14 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14216-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene, CCL14, is one of several CC cytokine genes clustered on 17q11.2. The CC cytokines are secreted proteins characterized by two adjacent cysteines. The cytokine encoded by this gene induces changes in intracellular calcium concentration and enzyme release in monocytes. This gene expresses both monocistronic and bicistronic transcripts. Bicistronic transcripts include the upstream cytokine gene CCL15. In addition, CCL14 undergoes alternative splicing in the bicistronic transcripts; it is unknown if its monocistronic transcript is also subject to alternative splicing.Spécification
CCL14 | |
Polyclonal | |
Unconjugated | |
CCL14 | |
C C motif chemokine 14, CC 1, CC 3, CCL14, Chemokine CC 1/CC 3, CKb1, HCC 1, HCC 1(1 74), HCC 1/HCC 3, HCC 3, MCIF, NCC 2, NCC2, SCYA14, SCYL2, Small inducible cytokine A14, SY14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6358 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16627 | |
CCL14 | |
CCL14 Fusion Protein Ag5439 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |