missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL17 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22342-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.Spécification
CCL17 | |
Polyclonal | |
Unconjugated | |
CCL17 | |
A 152E5.3, ABCD 2, C C motif chemokine 17, CC chemokine TARC, CCL17, SCYA17, Small inducible cytokine A17, TARC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6361 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92583 | |
CCL17 | |
CCL17 Fusion Protein Ag17832 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |