missing translation for 'onlineSavingsMsg'
Learn More

CCP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14067-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802544

  • 329.85€ / 150µL

Date d’expédition estimée: 21-06-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

NNA1 is a zinc carboxypeptidase that contains nuclear localization signals and an ATP/GTP-binding motif that was initially cloned from regenerating spinal cord neurons of the mouse.
Spécification

Spécification

CCP1
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q641K1, Q9UPW5
AGTPBP1, ATP/GTP binding protein 1, CCP1, Cytosolic carboxypeptidase 1, KIAA1035, NNA1
Rabbit
IgG
150 μL
-20°C
Primary
23287, 67269
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
AGTPBP1
Liquid
Agtpbp1
CCP1 Fusion Protein Ag5122
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Vidéos
FDS
Documents
Certificats