missing translation for 'onlineSavingsMsg'
Learn More

CD107a (LAMP-1) Rat anti-Mouse, eFluor® 660, Clone: eBio1D4B (1D4B), eBioscience™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 50-1071-80

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.25000kg

 Afficher plus de versions de ce produit

Code produit. 15331880

  • 94.00€ / 25µg

Expédition estimée
29-09-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. CD107a (lysosome-associated membrane protein-1, LAMP-1), together with LAMP-2, is a major constituent of lysosomal membrane; 1-2% of total CD107a is found also on the plasma membrane. The LAMP proteins are involved in lysosome biogenesis and are required for fusion of lysosomes with phagosomes. Increased CD107a immunoreactivity is observed in neurons, and in glial cells surrounding senile plaques in Alzheimer′s disease cases and is localized mainly in medullary epithelial cells, single macrophages and lymphocytes in acute thymic involution. CD107a is a good marker of mast cell activation.
Spécification

Spécification

CD107a (LAMP-1)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P11438
CD107a, LAMPA, LGP120, CD107 antigen-like family member A, LAMP-1, lysosome-associated membrane glycoprotein 1, lysosome-associated membrane protein 1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
eBio1D4B (1D4B)
eFluor™ 660
Lamp1
Liquid
Lamp1
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16783
Vidéos
Documents

For Research Use Only.