missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 (c-Kit) Monoclonal Antibody (104D2), APC-eFluor™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
265.00€ - 600.00€
Spécification
Antigène | CD117 (c-Kit) |
---|---|
Clone | 104D2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16636364
|
Affymetrix eBioscience
47-1178-41 |
25 Tests |
265.00€
25 pièces |
Expédition estimée: 16-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16646364
|
Affymetrix eBioscience
47-1178-42 |
100 Tests |
600.00€
100 pièces |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The 104D2 MAb reacts with human CD117, also known as c-Kit, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145 kDa molecule is expressed by hematopoietic progenitor cell subsets and mast cells. The interaction of c-Kit and Steel factor promotes proliferation and differentiation of hematopoietic progenitor cells and mast cell differentiation. CD117 is also expressed by melanocytes and plays a role in signaling and activation of these cells.
CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Human samples. It is validated for Flow Cytometry.Spécification
CD117 (c-Kit) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
3815 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
KIT |
104D2 | |
Flow Cytometry | |
APC-eFluor 780 | |
Mouse | |
Human | |
P10721 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |