missing translation for 'onlineSavingsMsg'
Learn More

CD117 (c-Kit) Mouse anti-Human, APC-eFluor 780, Clone: 104D2, eBioscience™ Invitrogen™

Mouse Monoclonal Antibody

Marque:  Invitrogen 47-1178-42

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16646364

  • 499.00€ / 100 pièces

Date d’expédition estimée: 09-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The 104D2 MAb reacts with human CD117, also known as c-Kit, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145 kDa molecule is expressed by hematopoietic progenitor cell subsets and mast cells. The interaction of c-Kit and Steel factor promotes proliferation and differentiation of hematopoietic progenitor cells and mast cell differentiation. CD117 is also expressed by melanocytes and plays a role in signaling and activation of these cells.

CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Human samples. It is validated for Flow Cytometry
Spécification

Spécification

CD117 (c-Kit)
Flow Cytometry
104D2
APC-eFluor 780
KIT
belly-spot; Bs; CD117; ckit; C-Kit; c-KIT gene1; C-KIT protein; c-kit proto-oncogene protein; c-kit receptor; c-kit receptor tyrosine kinase; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; kit oncogene; KIT proto-oncogene receptor tyrosine kinase; KIT proto-oncogene, receptor tyrosine kinase; mast cell growth factor receptor; mast/stem cell growth factor receptor; Mast/stem cell growth factor receptor Kit; MGF; p145 c-kit; PBT; Piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; SCFR; SCO1; SCO5; Sl; soluble KIT variant 1; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; Tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein; W
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3815
Liquid
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P10721
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Vidéos
FDS
Documents
Certificats