missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 Rat anti-Mouse, BB700, Clone: 2B8, BD Horizon™
Rat Monoclonal Antibody
Marque: BD Biosciences 566414
Informations supplémentaires : Poids : 0.01000kg
Description
CD117 Monoclonal antibody specifically detects CD117 in Mouse samples. It is validated for Flow Cytometry.Spécification
CD117 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
2B8 | |
Brilliant Blue 700 (BB700) | |
0.1 mg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |