missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD123 Mouse anti-Human, BB700, Clone: 7G3, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566483
Informations supplémentaires : Poids : 0.01000kg
Description
CD123 Monoclonal antibody specifically detects CD123 in Human samples. It is validated for Flow Cytometry.Spécification
CD123 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
25 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Flow Cytometry | |
7G3 | |
Mouse | |
RUO | |
Human | |
Purified |