Learn More
CD127 Mouse, anti-Human, R718, Clone: HIL-7R-M21, BD Horizon™
Mouse Monoclonal Antibody
Marque:  BD Biosciences 566967
Informations supplémentaires : Poids : 0.00060kg
 Afficher plus de versions de ce produit
Code produit. 17267754
  • 488.00€ / 100 tests
Expédition estimée: 02-01-2023
pour voir le stock disponible 
Description
Description
The hIL-7R-M21 monoclonal antibody specifically binds to the 60-90 kDa glycoprotein, CD127. CD127 is also known as the IL-7 receptor alpha (IL-7Rα) subunit. The IL-7 receptor complex is a heterodimer composed of CD127 and the common gamma chain (γc, CD132), shared by other cytokine receptors (IL-2R, IL-4R, IL-9R, IL-15R, and IL-21R). CD127 is expressed on thymocytes, T- and B-cell progenitors, mature T cells, and some lymphoid and myeloid cells. In vitro experiments show the expression of CD127 is down-regulated following T cell activation. Studies indicate that the IL-7 Receptor plays an important role in the proliferation and differentiation of mature T cells. Recently, it has been shown that low surface expression of CD127, in combination with intermediate to high surface expression of CD25, the α chain of the IL-2 receptor complex, can distinguish between human regulatory and conventional CD4+ T cells in human adult and cord blood, lymph nodes and thymus.
Spécification
Spécification
CD127
Monoclonal
R718
100 Tests
Primary
Refrigerate Upon Arrival
Flow Cytometry
HIL-7R-M21
Mouse