Learn More
Epredia™ Lab Vision™ CD138 Rabbit Polyclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD138, Rabbit Polyclonal Antibody.
142.80€ - 530.40€
Spécification
Antigène | CD138 |
---|---|
Concentration | 200µg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
12643587
|
Epredia™
RB-9422-P0 |
100 μL |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
12653587
|
Epredia™
RB-9422-P1 |
500 μL |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
12633587
|
Epredia™
RB-9422-P |
1 mL |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
CD138/Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence is a useful marker for viable myeloma cells.Host Species: Rabbit
Species Reactivity: Human. Others not tested
Epitope: aa 18-218
Immunogen: Recombinant protein encoding aa 18-218 human CD138
Molecular Weight: 30.5kDa (core protein); 85-92kDa (depending upon glycosaminoglycans)
Positive Control: Tonsil
Cellular Localization: Plasma cell and Membrane
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Spécification
CD138 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Adhesion | |
Recombinant protein encoding aa 18-218 human CD138 |
200µg/mL | |
Polyclonal | |
Concentrated | |
RUO | |
Human | |
Primary |