Learn More
CD14 Mouse anti-Baboon, Cynomolgus, Human, Rhesus, Brilliant Ultraviolet 737, Clone: M5E2, BD Horizon™
Mouse Monoclonal Antibody
446.00€ - 815.00€
Spécification
Antigène | CD14 |
---|---|
Clone | M5E2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | Brilliant Ultraviolet 737 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17267864
|
BD Biosciences
612764 |
25 Tests |
446.00€
25 tests |
Expédition estimée: 16-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17257864
|
BD Biosciences
612763 |
100 Tests |
815.00€
100 tests |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
The antibody was conjugated to BD Horizon™ BUV737 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 737-nm. BD Horizon Brilliant BUV737 can be excited by the ultraviolet laser (355 nm) and detected with a 740/35 filter. Due to the excitation of the acceptor dye by other laser lines, there may be significant spillover into channels detecting Alexa Fluor™ 700-like dyes (e.g., 712/20-nm filter).
Spécification
CD14 | |
Flow Cytometry | |
Brilliant Ultraviolet 737 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
M5E2 | |
Monoclonal | |
Mouse | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
Human CD14 Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |