missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, Non-human Primate, NovaFluor™ Red 660, Clone: MEM-15, Invitrogen™

Mouse Monoclonal Antibody

Marque:  Invitrogen H007T02R01

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 17266546

  • 158.00€ / 25 pièces
Expédition estimée: 29-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The MEM-15 monoclonal antibody reacts with human CD14, a 53-55 kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.
Applications Reported: This MEM-15 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This MEM-15 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 4 μL (0.6 μg) per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes.
When using more than one NovaFluor dye, CellBlox Monocyte and Macrophage Blocking Buffer may be used to block non-specific binding of dyes to monocytes.
Excitation: 637 nm; Emission: 659 nm; Laser: 633-640 nm (Red) Laser

CD14 Monoclonal antibody specifically detects CD14 in Human, Non-human Primate samples. It is validated for Flow Cytometry
Spécification

Spécification

CD14
Monoclonal
4 μL/Test
PBS with no preservative; pH 7.2
P08571
Mouse
25 Tests
Primary
Non-human Primate, Human
Antibody
IgG1
Flow Cytometry
MEM-15
NovaFluor Red 660
Cd14
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; Monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; Myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14
Affinity chromatography
RUO
929
4° C, store in dark, DO NOT FREEZE!
Liquid
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions