missing translation for 'onlineSavingsMsg'
Learn More

CD162 (PSGL-1) Monoclonal Antibody (4RA10), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 53-1621-82

288.68 EUR valable jusqu'au 2024-03-29
Utilisez le code promo "21615" pour bénéficier de cette offre.



Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16626404

  • 433.00€ / 1 pièce
En stock
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Description: This 4RA10 monoclonal antibody reacts with mouse CD162, which is also known as P-selectin glycoprotein ligand-1 (PSGL-1). This clone has been successfully used in flow cytometry and has been reported to block binding of CD162 to CD62P and CD62L. Applications Reported: This 4RA10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4RA10 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser

CD162 (PSGL-1) Monoclonal antibody specifically detects CD162 (PSGL-1) in Mouse samples. It is validated for Flow Cytometry.
Spécification

Spécification

CD162 (PSGL-1)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q62170
SELPLG
Mouse PSGL-1 human IgG1 fusion protein
100 μg
Primary
Mouse
Antibody
IgG1 κ
Flow Cytometry
4RA10
Alexa Fluor 488
SELPLG
CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG
Rat
Affinity chromatography
RUO
20345
4° C, store in dark, DO NOT FREEZE!
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions