missing translation for 'onlineSavingsMsg'
Learn More

CD19 Mouse anti-Human, APC-R700, Clone: HIB19, BD Horizon™

Mouse Monoclonal Antibody

Marque:  BD Biosciences 564977

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15873999

  • 610.00€ / 1 pièce
Expédition estimée: 23-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow cytometry.

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow cytometry.
Spécification

Spécification

CD19
Monoclonal
APC-R700
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Mouse
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
IgG1 κ
Flow Cytometry
HIB19
Vol. Per test: 5μL
B4; B-lymphocyte antigen CD19; Leu-12
100 Tests
Cell Biology
Human
Purified
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.