Learn More
CD193 (CCR3) Monoclonal Antibody (eBio5E8-G9-B4 (5E8-G9-B4)), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
267.00€ - 548.00€
Spécification
Antigène | CD193 (CCR3) |
---|---|
Clone | eBio5E8-G9-B4 (5E8-G9-B4) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16636444
|
Affymetrix eBioscience
56-1939-41 |
25 Tests |
267.00€
25 pièces |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16646444
|
Affymetrix eBioscience
56-1939-42 |
100 Tests |
548.00€
100 pièces |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The eBio5E8-G9-B4 MAb reacts with human CD193 (CCR3, CKR3). CD193 is a member of the seven transmembrane G-protein coupled receptor (GPCR) family, and is a high affinity chemokine receptor for the chemokines eotaxin-1 (CCL11), eotaxin-2 (CCL24), eotaxin-3 (CCL26) and MCP-4 (CCL13), but has also been reported to bind RANTES, MCP-3 and MCP-4. CD193 is highly expressed on the surface of eosinophils, and is the key eosinophil chemokine receptor responsible for the regulation of eosinophil migration and function. In addition to eosinophils, CD193 is also expressed on CD4+ TH2 cells, basophils, mast cells, mononuclear phagocytes, platelets, CD34+ hematopoietic progenitors and airway epithelial cells. CD193 likely plays an important role in the migration of eosinophils in allergic airway inflammation and asthma. CD193 can also serve as a receptor for HIV-1 cell entry.
CD193 (CCR3) Monoclonal antibody specifically detects CD193 (CCR3) in Human samples. It is validated for Flow Cytometry.Spécification
CD193 (CCR3) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
b-chemokine receptor; CC chemokine receptor 3; C-C chemokine receptor type 3; C-C CKR-3; C-C motif chemokine receptor 3; CC-CKR3; CC-CKR-3; Ccr3; CCR-3; CD193; chemokine (C-C motif) receptor 3; chemokine (C-C) receptor 1,-like 2; chemokine (C-C) receptor 3; CKR3; Cmkbr1l2; CMKBR3; eosinophil CC chemokine receptor 3; eosinophil eotaxin receptor; Macrophage inflammatory protein 1-alpha receptor-like 2; MGC102841; MIP-1 alpha RL2; MIP-1 alphaRL2; probable C-C chemokine receptor type 3 | |
CCR3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CCR3 |
eBio5E8-G9-B4 (5E8-G9-B4) | |
Flow Cytometry | |
Alexa Fluor 700 | |
Mouse | |
Human | |
P51677 | |
1232 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |