missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Rat anti-Mouse, BB515, Clone: 1B1, BD Horizon™
Rat Monoclonal Antibody
191.00€ - 427.00€
Spécification
Antigène | CD1d |
---|---|
Clone | 1B1 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15864629
|
BD Biosciences
565956 |
25 μg |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
15854629
|
BD Biosciences
565955 |
0.1 mg |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
CD1d Monoclonal antibody specifically detects CD1d in Mouse samples. It is validated for Flow cytometry.
CD1d Monoclonal antibody specifically detects CD1d in Mouse samples. It is validated for Flow cytometry.Spécification
CD1d | |
0.2mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Cd1d1; Cd1.1; Cd1a; Cd1d; Ly-38 | |
Primary |
1B1 | |
Flow Cytometry | |
Brilliant Blue 515 (BB515) | |
Rat | |
Cell Biology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Cd1.1 cDNA-transfected RMA-S mouse T lymphoma and L929 cells | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. |
For Research Use Only.