missing translation for 'onlineSavingsMsg'
Learn More

CD206 (MMR) Rat anti-Mouse, PerCP-eFluor™ 710, Clone: MR6F3, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marque:  Invitrogen 46-2061-80

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16370854

  • 131.00€ / 25µg

Expédition estimée
16-11-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD206 (MMR) Monoclonal antibody specifically detects CD206 (MMR) in Mouse samples. It is validated for Flow Cytometry
Spécification

Spécification

CD206 (MMR)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q61830
AW259686; bA541I19.1; CD206; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; hMR; Human mannose receptor; Macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; MMR; MR; Mrc1; MRC1L1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
MR6F3
PerCP-eFluor 710
Mrc1
Liquid
Mrc1
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
17533
Vidéos
Documents