missing translation for 'onlineSavingsMsg'
Learn More

CD21/CD35 Rat anti-Mouse, APC-eFluor® 780, Clone: eBio8D9 (8D9), eBioscience™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 47-0211-80

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.25000kg

 Afficher plus de versions de ce produit

Code produit. 15341430

  • 112.00€ / 25µg

Expédition estimée
30-10-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. CD21 (complement receptor 2, CR2) binds C3 complement fragments, especially its breakdown fragments, which remain covalently attached to complement activating surfaces or antigen. CD21 has important roles in uptake and retention of immunocomplexes, survival of memory B cells and in development and maintece of the humoral response to T-dependent antigens. CD21 also serves as a key receptor for Epstein-Barr virus binding and is involved in targeting prions to folicular dendritic cells and expediting neuroinvasion following peripheral exposure to prions. A soluble form of the CD21 (sCD21) is shed from the lymphocyte surface and retains its ability to bind respective ligands. CD35 encodes a membrane glycoprotein found on peripheral blood cells, glomerular podocytes, and follicular dendritic cells. The protein is a receptor for complement components C3b and C4b and regulates the activity of the complement cascade. Variation in this protein is the basis of the Knops blood group system. The two most common alleles, F and S, differ by 8 exons and are thought to be the result of an unequal crossover event. A secreted form of the protein present in plasma has been described, but its full length nature has not been determined.
Spécification

Spécification

CD21/CD35
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P19070
EBV receptor, complement C3d receptor, complement receptor type 2, epstein-Barr virus receptor, C3-binding protein, C3b/C4b receptor, CD35 antigen, Knops blood group antigen, complement component receptor 1, complement receptor type 1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
eBio8D9 (8D9)
APC-eFluor™ 780
Cr2
Liquid
Cr2
IgG2a, lambda
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12902
Vidéos
Documents

For Research Use Only.