missing translation for 'onlineSavingsMsg'
Learn More

CD23 Mouse anti-Human, Mouse, Clone: 4C7C9, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 60208-2-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837215

  • 325.38€ / 150µL
Expédition estimée: 10-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.
Spécification

Spécification

CD23
Monoclonal
0.9 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06734, P20693
Fcer2
CD23 Fusion Protein Ag0425
150 μL
Primary
Mouse, Human
Antibody
IgG2a
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
4C7C9
Unconjugated
FCER2
BLAST 2, CD23, CD23A, CLEC4J, Fc epsilon RII, FCE2, FCER2, IGEBF, Lymphocyte IgE receptor
Mouse
Protein A
RUO
14128, 2208
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions