Learn More
CD25 Mouse, PE, Clone: BC96, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio QAB35-PE-25Tests
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15981878
  • 94.25€ / 25 tests
Expédition estimée: 26-01-2024
Spécification
CD25
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
IL2RA
Protein A/G Purified
RUO
3559
IgG1 κ
Flow Cytometry
BC96
PE
NP_001295171
Mouse
25 Tests
Primary
Store at 2-4°C.