Learn More
CD27 Monoclonal Antibody (O323), Super Bright™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
152.00€ - 352.00€
Spécification
Antigène | CD27 |
---|---|
Clone | O323 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15964144
|
Affymetrix eBioscience
78-0279-42 |
100 Tests |
352.00€
100 pièces |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15954144
|
Affymetrix eBioscience
78-0279-41 |
25 Tests |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
Description: This HM36 antibody recognizes mouse CD36, also known as scavenger receptor BIII. Applications Reported: This HM36 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HM36 antibody has been tested by flow cytometric analysis of thioglycolate-elicited peritoneal macrophages. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. In some experiments, we have observed that compensation values for Super Bright 780-conjugated antibodies are higher in the violet 450/50 channel when using UltraComp eBeads microspheres (Product No. 01-2222-42) as compared to single-color stained cells. In such circumstances, we would recommend setting compensation with cells. We have also observed this in some experiments using AbC Total Antibody Compensation beads (Product No. A10497).
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry.Spécification
CD27 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P26842 | |
CD27 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD27 |
O323 | |
Flow Cytometry | |
Super Bright 780 | |
Mouse | |
Human | |
939 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |