Learn More
CD28 Mouse anti-Human, PE-Cy5, Clone: CD28.2, BD
Mouse Monoclonal Antibody
240.00€ - 545.00€
Spécification
Antigène CD28
Clone CD28.2
Applications Flow Cytometry
Classification Monoclonal
Conjugué PE-Cyanine5
Consulter d'autres spécifications
Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15861339
Afficher les documents Détails de la promotion
BD Biosciences
561791
25 Tests
240.00€
1 pièce
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 23-04-2024
pour voir le stock disponible 
15848398
Afficher les documents Détails de la promotion
BD Biosciences
555730
100 Tests
545.00€
1 pièce
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 23-04-2024
Description
The CD28.2 monoclonal antibody specifically binds to CD28, a 44 kDa homodimeric transmembrane glycoprotein present on most mature T cells, thymocytes and plasma cells. CD28 is a costimulatory receptor that binds CD80 and CD86 as ligands and plays a very important role in T cell-B cell interactions. It has been suggested that CD28 initiates and regulates a separate and distinct signal transduction pathway from those stimulated by the TCR complex. Additionally, it has been reported that CD28 antibody clones vary in their ability to stimulate T cells to produce IL-2 and increase intracellular Ca2+ concentration. This finding suggests the existence of functionally distinct subregions on the CD28 molecule. CD28.2 has been demonstrated to bind to the same molecule as clone L293, another CD28 mAb, and has been reported to induce Ca2+ influx in Jurkat T cells.
Spécification
Spécification
CD28
Flow Cytometry
PE-Cyanine5
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
CD28 antigen; T44; Tp44; TP44
CD28.2
Monoclonal
Mouse
Human
Human CD28 Transfected Cell Line
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Documents
Product Certifications
For Research Use Only.