missing translation for 'onlineSavingsMsg'
Learn More

CD304 (Neuropilin-1) Monoclonal Antibody (3DS304M), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

170.00€ - 414.00€

Spécification

Antigène CD304 (Neuropilin-1)
Clone 3DS304M
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15509236
Afficher les documents Détails de la promotion
Affymetrix eBioscience
46-3041-80
25 μg
170.00€
25µg
Voir les produits de remplacement
Économisez jusqu'à 
»
En stock
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15519236
Afficher les documents Détails de la promotion
Affymetrix eBioscience
46-3041-82
100 μg
414.00€
100µg
Voir les produits de remplacement
Économisez jusqu'à 
»
En stock
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Description: The monoclonal antibody 3DS304M recognizes mouse CD304 (Neuropilin-1), a type 1 transmembrane glycoprotein present on the surface of multiple cell types, including: regulatory T cells (Treg), NKT cells, DC, certain types of stem cells, neurons, endothelial cells, and some neoplastic cells. Neuropilin-1, in complex with plexins, serves as a co-receptor for type 3 semaphorins in growing neurons. It is also involved in the process of angiogenesis being a part of a functional receptor for VEGF in endothelial cells. In mice, Neuropilin-1 is expressed on thymus-derived natural Treg but not peripherally induced Treg. In addition, transient expression on recently activated non-regulatory T cells has been observed. Neuropilin-1 has been used as a marker of recent thymic emigrants in the mouse iNKT cell population. It has been shown that Neuropilin-1 forms a complex with TGF beta receptors, activating the latent form of TGF beta (LAP-TGF beta 1) and augmenting canonical Smad2/3 signaling. This 3DS304M antibody will recognize formaldehyde-fixed as well as methanol-treated epitopes. Applications Reported: This 3DS304M antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3DS304M antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test.

CD304 (Neuropilin-1) Monoclonal antibody specifically detects CD304 (Neuropilin-1) in Mouse samples. It is validated for Flow Cytometry.
Spécification

Spécification

CD304 (Neuropilin-1)
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
A5 protein; BDCA4; C530029I03; CD304; Neuropilin; neuropilin 1; Neuropilin1; neuropilin-1; Neuropilin-1 precursor (A5 protein); NP1; NP-1; Npn1; NPN-1; NRP; NRP 1; Nrp1; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; vascular endothelial cell growth factor 165 receptor; VEGF165R
NRP1
Primary
4° C, store in dark, DO NOT FREEZE!
NRP1
3DS304M
Flow Cytometry
PerCP-eFluor 710
Rat
Mouse
P97333
18186
IgG2a κ
Affinity chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications

For Research Use Only.