missing translation for 'onlineSavingsMsg'
Learn More

CD321 (F11R) Mouse anti-Human, PE, Clone: CSTEM27, eBioscience™

Mouse Monoclonal Antibody

Marque:  Invitrogen 12-3219-41

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15875333

  • 125.00€ / 25 pièces
Date d’expédition estimée: 13-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This monoclonal antibody CSTEM27 reacts with human CD321 (F11R, JAM-1, JAM-A). Applications Reported: This CSTEM27 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM27 antibody has been pre-diluted and tested by flow cytometric analysis of human iPSCs. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, stem cells, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is involved in the maintece of epithelial and endothelial barriers, hematopoiesis, angiogenesis, and tumor proliferation, invasion, and metastasis.
Spécification

Spécification

CD321 (F11R)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9Y624
F11 R, JAMA, junctional adhesion molecule A, JAM-1, JAM-A
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
CSTEM27
PE
F11R
Liquid
F11R
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
50848
Vidéos
FDS
Documents
Certificats
Promotions

Promotions