Learn More
CD335 (NKp46) Monoclonal Antibody (9E2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
230.00€ - 444.00€
Spécification
Antigène | CD335 (NKp46) |
---|---|
Clone | 9E2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15835603
|
Affymetrix eBioscience
63-3359-41 |
25 Tests |
230.00€
25 pièces |
Expédition estimée: 22-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15845603
|
Affymetrix eBioscience
63-3359-42 |
100 Tests |
444.00€
100 pièces |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Description: This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes. The 9E2 antibody has been reported to induce cytotoxicity. Applications Reported: This 9E2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9E2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human periheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat.
CD335 (NKp46) Monoclonal antibody specifically detects CD335 (NKp46) in Human samples. It is validated for Flow Cytometry.Spécification
CD335 (NKp46) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
1-Mar; activating receptor 1; Ar1; CD335; hNKp46; Kilr1; KILR-1; LY94; Ly-94; lymphocyte antigen 94; Lymphocyte antigen 94 homolog; lymphocyte antigen 94 homolog (activating NK-receptor; lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46); mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; Ncr1; NK cell-activating receptor; NK receptor KILR-1; NKACTR; Nkp46; NK-p46; NK-p46); rAR-1; rat-activating receptor 1 | |
NCR1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
NCR1 |
9E2 | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Human | |
O76036 | |
9437 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only