Learn More
CD357 / GITR Rat, FITC, Clone: DTA-1, enQuireBio™
Rat Monoclonal Antibody
Marque:  EnquireBio QAB72-F-25ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15972408
  • 131.00€ / 25µg
Expédition estimée: 17-05-2024
Spécification
CD357 / GITR
Monoclonal
0.5mg/mL
10mM Tris, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH8
Tnfrsf18
Protein A/G Purified
RUO
21936
IgG2b
Flow Cytometry
DTA-1
FITC
NP_033426
Rat
25 μg
Primary
Store at 2-4°C.