missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Monoclonal Antibody (APA1/1), PE, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110183
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody recognizes an activation-dependent intracellular epitope of CD3 epsilon.
The CD3 subunit complex which is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. The genes encoding the CD3 epsilon, gamma and delta polypeptides are located on chromosome 11. Defects in the CD3 gene are associated with CD3 immunodeficiency.Spécification
CD3e | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P04234, P04235, P07766, P09693, P11942, P20963, P22646, P24161 | |
Cd247, Cd3d, CD3E, CD3g | |
Purified human CD3 proteins isolated from thymus | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Flow Cytometry | |
APA1/1 | |
PE | |
CD3g | |
4930549J05Rik; A430104F18Rik; AI504783; antigen CD3D, delta polypeptide (TiT3 complex); antigen CD3E, epsilon polypeptide (TiT3 complex); antigen CD3G, gamma polypeptide; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; CD3; CD3 antigen delta chain; CD3 antigen delta polypeptide; CD3 antigen gamma chain; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 epsilon chain; CD3 epsilon subunit; CD3 epsilon subunit precursor; CD3 gamma-chain; CD3 glycoprotein; CD3 glycoprotein precursor; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3 protein; CD3 TCR complex; CD3 type I transmembrane glycoprotein; CD3 type I transmembrane glycoprotein precursor; CD3 zeta chain; Cd3d; CD3D antigen d | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
12500, 12501, 12502, 12503, 915, 916, 917, 919 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |