Learn More
CD4 Mouse anti-Human, BB700, Clone: SK3 (also known as Leu3a), BD Horizon™
Mouse Monoclonal Antibody
Marque:  BD Biosciences 566392
Informations supplémentaires : Poids : 0.01000kg
 Afficher plus de versions de ce produit
Code produit. 15875009
  • 678.30€ / 1 pièce
Date d’expédition estimée: 22-12-2021
Description
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Spécification
Spécification
CD4
SK3 (also known as Leu3a)
Purified
Mouse
100 Tests
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
BB700
Mouse BALB/c IgG1, κ
IgG1, κ