Learn More
CD4 Monoclonal Antibody (OX35), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
130.00€ - 301.00€
Spécification
Antigène | CD4 |
---|---|
Clone | OX35 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | PerCP-eFluor 710 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15341220
|
Affymetrix eBioscience
46-0040-80 |
25 μg |
130.00€
25µg |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15351220
|
Affymetrix eBioscience
46-0040-82 |
100 μg |
301.00€
100µg |
Expédition estimée: 05-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Description: The OX35 monoclonal antibody reacts with rat CD4. CD4 is a co-receptor which participates in signaling through the T cell receptor by making contacts with MHC class II expressed on antigen-presenting cells (APC). CD4 is expressed in the thymus by CD4+CD8+ double-positive and CD4+ single-positive thymocytes, and by CD4+ helper T cells in the periphery. CD4 is a cell-surface membrane glycoprotein which contains four extracellular immunoglobulin-like domains. Applications Reported: This OX35 antibody has been reported for use in flow cytometric analysis. Applications Tested: This OX35 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.
CD4 Monoclonal antibody specifically detects CD4 in Rat samples. It is validated for Flow Cytometry.Spécification
CD4 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Rat | |
24932 | |
Primary | |
Antibody |
OX35 | |
Monoclonal | |
Liquid | |
RUO | |
P05540 | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.