missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.2 Mouse anti-Mouse, BB700, Clone: 104, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566497
Informations supplémentaires : Poids : 0.00810kg
Description
CD45.2 Monoclonal antibody specifically detects CD45.2 in Mouse samples. It is validated for Flow Cytometry.Spécification
CD45.2 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
104 | |
Brilliant Blue 700 (BB700) | |
25 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |