Learn More
CD45 Rat, Qfluor 710, Clone: 30-F11, enQuireBio™
Rat Monoclonal Antibody
Marque: EnquireBio QAB40-QF710-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD45 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
Ptprc | |
Protein A/G Purified | |
RUO | |
19264 |
Flow Cytometry | |
30-F11 | |
Qfluor 710 | |
NP_001255215 | |
Rat | |
25 μg | |
Primary | |
IgG2b κ |