missing translation for 'onlineSavingsMsg'
Learn More

CD47 Monoclonal Antibody (2D3), NovaFluor™ Blue 690, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marque:  Invitrogen H065T02B09

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 17896621

  • 155.00€ / 25 pièces
Expédition estimée: 17-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow Cytometry
Spécification

Spécification

CD47
Monoclonal
0.6 μg/Test
PBS with 0.09% sodium azide; pH 7.2
Q08722
Mouse
25 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
2D3
NovaFluor Blue 690
CD47
9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47
Affinity chromatography
RUO
961
4° C, store in dark, DO NOT FREEZE!
Liquid
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions