missing translation for 'onlineSavingsMsg'
Learn More

CD49b (Integrin alpha 2) Rat anti-Mouse, APC, Clone: DX5, eBioscience™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 17-5971-82

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.25000kg

 Afficher plus de versions de ce produit

Code produit. 15340810

  • 270.00€ / 100µg

En stock, prêt à être expédié : 4

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD49b (Integrin alpha 2, Integrin alpha 2 chain, VLA-2 alpha chain, HM alpha 2) is a member of the integrin family. It is a glycoprotein with molecular weight of 150 kD, and it complexes with CD29 (Integrin beta 1) to form the heterodimeric integrin VLA-2 (integrin alpha 2 beta 1, or GPIa/IIa) complex. VLA-2 is an extracellular receptor for laminin, collagen, and fibronectin, and interaction with its ligands results in the activation of intracellular signaling pathways. It has reported roles in VEGF-induced angiogenesis in vivo, as well as adhesion and lymphocyte activation. CD49b is expressed by NK cells, NK-T cells, monocytes, platelets, and epithelial cells. It is also expressed on adaptive immune cells such as T and B cells, specifically on a subset of CD4+ T cells in the spleen, on intraepithelial and lamina propria lymphocytes in the intestine, as well as on a population of peripheral CD4+ type 1 T regulatory (Tr1) cells that co-express LAG-3.
Spécification

Spécification

CD49b (Integrin alpha 2)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q62469
BR, CD49B, GPIa, VLA-2, VLAA2, ITGA2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
DX5
APC
Itga2
Liquid
Itga2
IgM, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16398
Vidéos
Documents

For Research Use Only.