Learn More
CD49d Mouse anti-Human, APC, Clone: 9F10, BD
Mouse Monoclonal Antibody
270.00€ - 585.00€
Spécification
Antigène | CD49d |
---|---|
Clone | 9F10 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | APC |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15871439
|
BD Biosciences
561892 |
25 Tests |
270.00€
1 pièce |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15889808
|
BD Biosciences
559881 |
100 Tests |
585.00€
1 pièce |
Expédition estimée: 14-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The 9F10 monoclonal antibody specifically reacts with the integrin α4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β1 (CD29) or β7. The α4β1 integrin (VLA-4) is expressed on lymphocytes, monocytes, thymocytes, NK cells, and several B- and T-cell lines, and mediates binding to VCAM-1 (CD106) and the CS-1 region of fibronectin. The αβ7 also binds fibronectin and VCAM-1, and has been shown in the mouse to preferentially bind the mucosal vascular addressin molecule, MAdCAM-1. This antibody is useful for studies of the expression by and function of cells that express α4 chain-containing integrins.
Spécification
CD49d | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
9F10 | |
Monoclonal | |
Mouse | |
Baboon, Bovine, Canine, Cynomolgus, Equine, Feline, Human, Rhesus Monkey, Sheep | |
IgG1 κ | |
Affinity Purified | |
Integrin α4 chain; Integrin alpha 4; ITGA4; IA4; alpha 4 subunit of VLA-4 |
For Research Use Only.