Learn More
CD53 Monoclonal Antibody (MEM-53), PE, Invitrogen™
Mouse Monoclonal Antibody
Marque:  Invitrogen MA110248
Code produit. 12382263
  • 450.00€ / 100 pièces
Découvrez plus d'offres spéciales
Description
Description
This antibody reacts with CD53, a 32-40 kDa glycoprotein, and also This antibody reacts with the deglycosylated molecule (molecular weight of the antigen is reduced by 15 kDa using endoglycosidase F).
CD53 is a tetraspanin family transmembrane glycoprotein expressed in the lymphoid-myeloid lineage. This molecule has been reported to form complexes with other leukocyte surface proteins such as CD2, CD19, CD21, MHC II, VLA-4 or tetraspanins CD37, CD81 and CD82, thus probably modulating various signaling processes. CD53 is involved in radioresistancy of tumor cells and its triggering has anti-apoptotic effect. In thymus, CD53 is up-regulated in response to positive selection signals during T cell development, and is strongly expressed upon macrophage exposure to bacterial lipopolysaccharide, whereas stimulation of neutrophils results in down-regulation of CD53 expression.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
CD53
Monoclonal
PE
Cd53
AI323659; antigen MOX44 identified by monoclonal antibody MRC-OX44; Cd53; CD53 antigen; CD53 glycoprotein; CD53 molecule; CD53 tetraspan antigen; cell surface antigen; cell surface glycoprotein CD53; Leukocyte antigen MRC OX-44; leukocyte surface antigen CD53; MOX44; OX44; Ox-44; tetraspanin-25; transmembrane glycoprotein; TSPAN25; tspan-25
Mouse
Size-exclusion chromatography
RUO
963
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MEM-53
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P19397
Cd53
Leukocytes of pacient suffering from a LGL-type leukemia.
100 Tests
Primary
Human
Antibody
IgG1