missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD58 Monoclonal Antibody (MEM-63), PE
Mouse Monoclonal Antibody
Marque: Invitrogen™ MA110256
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody reacts with CD58 (LFA-3), a 40-70 kDa extracellular membrane glycoprotein distributed over many tissues, leukocytes, erythrocytes, endothelial cells, epithelial cells and fibroblasts.
This gene encodes a member of the immunoglobulin superfamily. The encoded protein is a ligand of the T lymphocyte CD2 protein, and functions in adhesion and activation of T lymphocytes. The protein is localized to the plasma membrane. Alternatively spliced transcript variants have been described. [provided by RefSeq, Jan 2009].
Spécification
CD58 | |
Monoclonal | |
PE | |
CD58 | |
Ag3; CD305; CD58; CD58 antigen; CD58 antigen, (lymphocyte function-associated antigen 3); CD58 molecule; FLJ23181; FLJ43722; LAIR1; LAIR-1; LFA3; LFA-3; Lymphocyte function-associated antigen 3; membrane receptor; RP5-1086K13.2; surface glycoprotein LFA-3 | |
Mouse | |
Size-exclusion Chromatography | |
RUO | |
396702, 965 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-63 | |
TBS with 0.2% BSA and 15mM sodium azide; pH 8.0 | |
P19256 | |
CD58 | |
NALM-6 human pre-B cell line. | |
100 Tests | |
Primary | |
Human, Pig | |
Antibody | |
IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu