missing translation for 'onlineSavingsMsg'
Learn More

CD62L Mouse anti-Human, BV786, Clone: SK11, BD Horizon™

Mouse Monoclonal Antibody

Marque:  BD Biosciences 565311

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15874229

  • 635.00€ / 1 pièce
Expédition estimée: 22-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD62L Monoclonal antibody specifically detects CD62L in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.

CD62L Monoclonal antibody specifically detects CD62L in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Spécification

Spécification

CD62L
Monoclonal
Brilliant Violet 786
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Cell Biology
Baboon, Cynomolgus, Human, Rhesus Monkey
Purified
Flow Cytometry
SK11 (also known as Anti-Leu-8)
Vol. Per test: 5μL
L-selectin; LECAM-1; SELL; LNHR; LSEL; LAM-1; LEU8; PLNHR; TQ-1; MEL-14
Human Peripheral Blood T Lymphocytes
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed.
IgG2a κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.