missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD66 Monoclonal Antibody (CD66a-B1.1), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marque: Affymetrix eBioscience 62-0668-42
Informations supplémentaires : Poids : 0.01000kg
Description
Mouse Monoclonal Antibody
CD66 Monoclonal antibody specifically detects CD66 in Human samples. It is validated for Flow Cytometry.Spécification
CD66 | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
O75871, P06731, P13688, P40198, P40199 | |
CEACAM1, CEACAM3, CEACAM4, CEACAM5, CEACAM6 | |
Affinity chromatography | |
RUO | |
1048, 1084, 1089, 4680, 634 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
CD66a-B1.1 | |
Super Bright 436 | |
CEACAM5 | |
antigen CD66; BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen; Carcinoembryonic antigen CGM1; carcinoembryonic antigen CGM7; carcinoembryonic antigen gene family member 1; carcinoembryonic antigen gene family member 7; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen related cell adhesion molecule 4; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); carcinoembryonic antigen-related cell adhesion molecule 3; Carcinoembryonic antigen-related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 4-sv1; carcinoembryonic antigen-related cell adhesion molecule 4-sv2; carcinoembryonic antigen-related cell adhesion molecule 5; Carcinoembryonic | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a κ |