missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21063-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.Spécification
CD79B | |
Polyclonal | |
Unconjugated | |
CD79B | |
B29, CD79B, Ig beta, IGB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
974 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P40259 | |
CD79B | |
CD79B Fusion Protein Ag15086 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |