missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Monoclonal Antibody (EM-04), PE, Invitrogen™
Rat Monoclonal Antibody
Marque: Invitrogen MA110312
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD9 antigen is a glycoprotein expressed on the surface of developing B lymphocytes, platelets, monocytes, eosinophils, basophil, stimulated T lymphocytes and by neurons and glial cells in the peripheral nervous system. CD9 belongs to a family of membrane proteins termed tetraspanins which transverse the membrane four times. In pre B cells and platelets, CD9 antigen regulates cell activation and aggregation possibly through an association with the integrin CD41 / CD61 (GPIIb / GPIIIa). CD9 is involved in cell motility, osteoclastogenesis, neurite outgrowth, myotube formation, and sperm-egg fusion, plays roles in cell attachment and proliferation and is necessary for association of heterologous MHC II molecules on the dendritic cell plasma membrane which is important for effective T cell stimulation. CD9 functions in many cellular processes including differentiation, adhesion, and signal transduction, and expression plays a critical role in the suppression of cancer cell motility and metastasis. CD9 is also considered as metastasis suppressor in solid tumors.Spécification
CD9 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P40240 | |
CD9 | |
Permeabilized murine bone marrow-derived mast cells (BMMC). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry | |
EM-04 | |
PE | |
CD9 | |
5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29 | |
Rat | |
Size-exclusion chromatography | |
RUO | |
12527 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |