missing translation for 'onlineSavingsMsg'
Learn More

CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), Super Bright™ 600, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

140.00€ - 364.00€

Spécification

Antigène CD90.1 (Thy-1.1)
Clone HIS51
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15973524
Afficher les documents Détails de la promotion
Affymetrix eBioscience
63-0900-82
100 μg
364.00€
100µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 16-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15963524
Afficher les documents Détails de la promotion
Affymetrix eBioscience
63-0900-80
25 μg Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet
N/A Voir les produits de remplacement
Économisez jusqu'à 
»
Description

Description

Description: The HIS51 monoclonal antibody reacts with rat CD90 and cross-reacts with mouse CD90.1, a GPI-linked membrane molecule. In the rat, CD90 is expressed by hematopoietic stem cells, immature B cells, thymocytes, recent thymic emigrants, neurons, inflammed endothelia and other cell types. In the CD90.1-expressing mouse strains, PL and AKR, CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells. Applications Reported: This HIS51 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIS51 antibody has been tested by flow cytometric analysis of rat splenocytes. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions.

CD90.1 (Thy-1.1) Monoclonal antibody specifically detects CD90.1 (Thy-1.1) in Mouse, Rat samples. It is validated for Flow Cytometry.
Spécification

Spécification

CD90.1 (Thy-1.1)
0.2 mg/mL
Monoclonal
Liquid
RUO
P01830, P01831
Thy1
Primary
4° C, store in dark, DO NOT FREEZE!
Thy1
HIS51
Flow Cytometry
Super Bright 600
Mouse
Mouse, Rat
21838, 24832
IgG2a κ
Affinity chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications