missing translation for 'onlineSavingsMsg'
Learn More

CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), Super Bright™ 436, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 62-0902-82

228.67 EUR valable jusqu'au 2024-03-29
Utilisez le code promo "21615" pour bénéficier de cette offre.



Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15963394

  • 343.00€ / 100µg
Expédition estimée: 23-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Description: The 53-2.1 monoclonal antibody reacts with mouse CD90.2 also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 53-2.1. CD90 is involved in regulation of adhesion and signal transduction by T cells. Applications Reported: This 53-2.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 53-2.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.03 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.

CD90.2 (Thy-1.2) Monoclonal antibody specifically detects CD90.2 (Thy-1.2) in Mouse samples. It is validated for Flow Cytometry.
Spécification

Spécification

CD90.2 (Thy-1.2)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P01831
Thy1
Affinity chromatography
RUO
21838
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
53-2.1
Super Bright 436
Thy1
CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions