missing translation for 'onlineSavingsMsg'
Learn More

CD94 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13332-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831484

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Natural killer (NK) cells, like CD94, are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.
Spécification

Spécification

CD94
Polyclonal
Unconjugated
KLRD1
CD94, KLRD1, KP43, NK cell receptor
Rabbit
Antigen Affinity Chromatography
RUO
3824
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13241
KLRD1
CD94 Fusion Protein Ag4142
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions